Acetonitril 99,8 GC, corrected for water content, ULTRA RESI-ANALYZED® Organic_Residue_Analysis, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

Inert_Gas_filled

9255-02EA 289.34 EUR
9255-02
Acetonitril 99,8 GC, corrected for water content, ULTRA RESI-ANALYZED® Organic_Residue_Analysis, J.T.Baker®
Acetonitril
Formule: H₃CCN
MW: 41.05 g/mol
Kookpunt: 81.6
Smeltpunt: –45.7 °C
Dichtheid: 0.782 g/cm³ (20 °C)
Vlampunt: 2 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001878
CAS nummer: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
Bereik: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 5
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (CH₃CN) (by GC, corrected for water) ≥99.8 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.6
Water (by KF, coulometric) ≤0.05 %
MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR