Methyleenchloride, BAKER ANALYZED® HPLC HPLC, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Methylene chloride

9410.2500EA 258.63 EUR
9410.2500 9410.1000
Methyleenchloride, BAKER ANALYZED® HPLC HPLC, J.T.Baker®
Methyleenchloride
Formule: CH₂Cl₂
MW: 84.93 g/mol
Kookpunt: 39.8
Smeltpunt: –95 °C
Dichtheid: 1.322 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000881
CAS nummer: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) min. 99.8 %
Density (g/ml) at 25°C 1.315 - 1.321
Chloride (Cl) max. 10 ppm
Preservative (Amylene) (ppm) about 20
Residue after Evaporation (in ppm) max. 2
Titrable Acid (meq/g) max. 0.0003
Water (H2O) max. 0.02 %
Ultraviolet Absorbance (1.00-cm path vs water):
at 254 nm max. 0.01
at 280 nm max. 0.01
at 350 nm max. 0.01
UV Cut-off, nm max. 233


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR