Natriumhypochloriet 5% available chlorine in aqueous solution, BAKER ANALYZED®, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Management of Change (MOC) category = N

9416-03EA 409.34 EUR
9416-03 9416-01
Natriumhypochloriet 5% available chlorine in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Natriumhypochloriet
Formule: ClNaO
Kookpunt: 111 °C (1013 hPa)
Smeltpunt: –30…–20 °C
Dichtheid: 1,22 to 1.25 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011120
CAS nummer: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance (Clear, light yellow liquid) Passes Test
Available Chlorine (when packaged) 5.0 - 6.0 %
Phosphate (PO₄) ≤0.0005 %
Calcium (Ca) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR