Supelco® Organic Reference Standard, Pullulan Standard

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

97873-25MG
97873-25MGEA 365 EUR
97873-25MG
Supelco® Organic Reference Standard, Pullulan Standard
Standaarden Organische standaarden

Order Now

Specificaties

Environmentally Preferable
Shipping conditions STD
Storage conditions 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR