Milk, non fat (skimmed milk), powder

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A0830.0500 A0830.1000 A0830.5000
A0830.0500EA 32 EUR
A0830.0500 A0830.1000 A0830.5000
Milk, non fat (skimmed milk), powder
Milk, non fat (skim milk)
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 999999-99-4
EINECS: 310-127-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR