2-Tolualdehyde ≥98% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: o-Tolualdehyde, 2-Methylbenzaldehyde

Stabilisatie: Stabilised with hydroquinone (HQ)

A10172.14 A10172.22
A10172.14EA 41.2 EUR
A10172.14 A10172.22
2-Tolualdehyde ≥98% stabilised
2-Tolualdehyde

Stabilized with 0.1% hydroquinone


Purity: 98% CAS No.: 529-20-4 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 67°(152°F) Refractive Index: 1.5470 Odour: Almond-like Melting Point: -35° Boiling Point: 199-200° Beilstein Reference: 605841 Merck Reference: 14,9527 MDL No.: MFCD00003338

Formule: C₈H₈O
MW: 120.15 g/mol
Kookpunt: 199…200 °C
Smeltpunt: –35 °C
Dichtheid: 1.037
Vlampunt: 67 °C (152 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003338
CAS nummer: 529-20-4
EINECS: 208-452-2
Merck Index: 14,9527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR