Propargylalcohol ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A10295.0F A10295.AE A10295.AP
A10295.0FEA 197 EUR
A10295.0F A10295.AE A10295.AP
Propargylalcohol ≥99%
Propargylalcohol

Purity: 99% CAS No.: 107-19-7 Molecular Formula: C3H4O Formula Weight: 56.06 Storage Temperature: 2°C to 8°C Flash Point: 33°(91°F) Refractive Index: 1.4320 Melting Point: -53° Boiling Point: 114-115° Beilstein Reference: 506003 Merck Reference: 14,7809 UN No.: 2929 MDL No.: MFCD00002912

Formule: C₃H₄O
Kookpunt: 114…115 °C
Smeltpunt: –53 °C
Dichtheid: 0,949
Vlampunt: 33 °C (91 °F)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002912
CAS nummer: 107-19-7
EINECS: 203-471-2
UN: 2929
ADR: 6.1,II
Merck Index: 14,7809

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR