2-Chloorfenol ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-Chloro-2-hydroxybenzene

A10311.30 A10311.0B A10311.0I
A10311.30EA 44.7 EUR
A10311.30 A10311.0B A10311.0I
2-Chloorfenol ≥99%
2-Chloorfenol

Purity: 99% CAS No.: 95-57-8 Molecular Formula: C6H5ClO Formula Weight: 128.56 Flash Point: 63°(145°F) Refractive Index: 1.5590 Odour: Phenol-like Melting Point: 7-8° Boiling Point: 175-176° Beilstein Reference: 1905114 Merck Reference: 14,2154 UN No.: 2021 MDL No.: MFCD00002159

Formule: C₆H₅ClO
MW: 128.56 g/mol
Kookpunt: 175…176 °C
Smeltpunt: 7…8 °C
Dichtheid: 1.260
Vlampunt: 63 °C (145 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002159
CAS nummer: 95-57-8
EINECS: 202-433-2
UN: 2021
ADR: 6.1 III
Merck Index: 12,02173

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR