Triethylsilane ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A10320.18 A10320.09 A10320.30
A10320.18EA 80.7 EUR
A10320.18 A10320.09 A10320.30
Triethylsilane ≥98%
Triethylsilane

Purity: 98+% CAS No.: 617-86-7 Molecular Formula: C6H16Si Formula Weight: 116.28 Flash Point: -6°(21°F) Refractive Index: 1.4120 Melting Point: -157° Boiling Point: 107-108° Beilstein Reference: 1098278 UN No.: 1993 MDL No.: MFCD00009018

Formule: C₆H₁₆Si
Kookpunt: 107…108 °C
Smeltpunt: –157 °C
Dichtheid: 0,728
Vlampunt: –6 °C (21 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009018
CAS nummer: 617-86-7
EINECS: 210-535-3
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR