Ninhydrin ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate

A10409.09 A10409.14 A10409.18 A10409.30
A10409.09EA 29.8 EUR
A10409.09 A10409.14 A10409.18 A10409.30
Ninhydrin ≥99%
Ninhydrin

Purity: 99% CAS No.: 485-47-2 Molecular Formula: C9H6O4 Formula Weight: 178.14 Melting Point: 251° dec. Beilstein Reference: 1910963 Merck Reference: 14,6554 MDL No.: MFCD00003791

Formule: C₉H₆O₄
MW: 178.14 g/mol
Kookpunt: 351
Smeltpunt: 251 °C (ontbinding)
Dichtheid: 0.862 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003791
CAS nummer: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR