Aminoacetaldehyde diethyl acetal 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal

A10427.14 A10427.22
A10427.14EA 64.8 EUR
A10427.14 A10427.22
Aminoacetaldehyde diethyl acetal 98%
Aminoacetaldehyde diethyl acetal

Purity: 98% CAS No.: 645-36-3 Molecular Formula: C6H15NO2 Formula Weight: 133.19 Flash Point: 45°(113°F) Refractive Index: 1.4170 Boiling Point: 162-163° Beilstein Reference: 506067 UN No.: 2733 MDL No.: MFCD00008136

Formule: C₆H₁₅NO₂
MW: 133.19 g/mol
Kookpunt: 162…163 °C
Dichtheid: 0.916
Vlampunt: 45 °C (113 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008136
CAS nummer: 645-36-3
EINECS: 211-439-4
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR