Benzamide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Benzoic acid amide

A10501.0E A10501.22 A10501.36
A10501.0EEA 236 EUR
A10501.0E A10501.22 A10501.36
Benzamide ≥98%
Benzamide

Purity: 98+% CAS No.: 55-21-0 Molecular Formula: C7H7NO Formula Weight: 121.14 Flash Point: 180°(356°F) Melting Point: 125-129° Boiling Point: 288° Beilstein Reference: 385876 Merck Reference: 14,1060 MDL No.: MFCD00007968

Formule: C₇H₇NO
MW: 121.14 g/mol
Kookpunt: 288 °C
Smeltpunt: 125…129 °C
Dichtheid: 1.341
Vlampunt: 180 °C (356 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007968
CAS nummer: 55-21-0
EINECS: 200-227-7
Merck Index: 13,01059

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR