1-Bromononane ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A10577.14 A10577.22 A10577.36
A10577.14EA 34 EUR
A10577.14 A10577.22 A10577.36
1-Bromononane ≥99%
1-Bromononane

Purity: 99% CAS No.: 693-58-3 Molecular Formula: CH3(CH2)8Br Formula Weight: 207.16 Flash Point: 90°(194°F) Refractive Index: 1.4550 Boiling Point: 200-201° Beilstein Reference: 1733632 MDL No.: MFCD00000278

Formule: BrH₂C(CH₂)₇CH₃
Kookpunt: 200…201 °C
Smeltpunt: –29 °C
Dichtheid: 1,086
Vlampunt: 90 °C (194 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000278
CAS nummer: 693-58-3
EINECS: 211-755-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR