L(+)-Diethyl tartrate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (+)-Diethyl L-tartrate, (R,R)-(+)-Tartaric acid diethyl ester, (2R,3R)-(+)-Diethyl tartrate, Diethyl L-tartrate, L(+)-Tartaric acid diethyl ester, (2R,3R)-Diethyl 2,3-dihydroxysuccinate, (R,R)-(+)-Diethyl tartrate

A10641.0B A10641.22 A10641.30 A10641.36
A10641.0BEA 196 EUR
A10641.0B A10641.22 A10641.30 A10641.36
L(+)-Diethyl tartrate ≥98%
L(+)-Diethyl tartrate

Purity: 98% CAS No.: 87-91-2 Molecular Formula: C8H14O6 Formula Weight: 206.19 Flash Point: 93°(199°F) Refractive Index: 1.4460 Melting Point: 17° Boiling Point: 280° Beilstein Reference: 1727145 Merck Reference: 14,3855 Optical Rotation: +27° (c=1 in water) MDL No.: MFCD00009143

Formule: C₈H₁₄O₆
MW: 206.2 g/mol
Kookpunt: 280 °C
Smeltpunt: 17 °C
Dichtheid: 1.205
Vlampunt: 93 °C (199 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009143
CAS nummer: 87-91-2
EINECS: 201-783-3
Merck Index: 13,03886

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR