D-(+)-Camphor ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A10708.14 A10708.22 A10708.36
A10708.14EA 25.6 EUR
A10708.14 A10708.22 A10708.36
D-(+)-Camphor ≥98%
D-(+)-Camphor

Purity: 98% CAS No.: 464-49-3 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 65°(149°F) Odour: Camphor-like Boiling Point: 204° Beilstein Reference: 2042745 Merck Reference: 14,1732 Optical Rotation: +43° (c=10 in ethanol) UN No.: 2717 MDL No.: MFCD00064149

Formule: C₁₀H₁₆O
Kookpunt: 204 °C
Smeltpunt: 175…180 °C
Dichtheid: 0,990
Vlampunt: 65 °C (149 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064149
CAS nummer: 464-49-3
EINECS: 207-355-2
UN: 2717
ADR: 4.1,III
Merck Index: 14,1732

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR