3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A10732.09 A10732.18 A10732.30
A10732.09EA 51 EUR
A10732.09 A10732.18 A10732.30
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt ≥99%
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt

Purity: 99% CAS No.: 54970-72-8 Molecular Formula: C6H3Cl2NaO4S Formula Weight: 265.05 Melting Point: 300° Beilstein Reference: 3640643

Formule: C₆H₃Cl₂NaO₄S
Smeltpunt: >300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009798
CAS nummer: 54970-72-8
EINECS: 259-416-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR