γ-(N-Morpholino)propanesulphonic acid (MOPS)

Leverancier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

A1076.0250 A1076.1000 A1076.5000
A1076.0250EA 146 EUR
A1076.0250 A1076.1000 A1076.5000
γ-(N-Morpholino)propanesulphonic acid (MOPS)
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formule: C₇H₁₅NO₄S
MW: 209.27 g/mol
Smeltpunt: > 250 °C
Vlampunt: 110 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006183
CAS nummer: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR