Hydrazinium sulphate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A10792.30 A10792.A1
A10792.30EA 33.2 EUR
A10792.30 A10792.A1
Hydrazinium sulphate ≥99%
Hydrazinium sulphate

Purity: 99+% CAS No.: 10034-93-2 Molecular Formula: H2NNH2.H2SO4 Formula Weight: 130.12 Odour: Odourless pH: 1.3 Melting Point: 254° Merck Reference: 14,4772 UN No.: 3288 MDL No.: MFCD00044873

Formule: NH₂NH₂·H₂SO₄
Smeltpunt: 254 °C
Dichtheid: 1,370
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00044873
CAS nummer: 10034-93-2
EINECS: 233-110-4
UN: 3288
ADR: 6.1,III
Merck Index: 14,4772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e277940036b3cf71839ff7458f0c7d08bff6f6d5&vid=7a677e6f1725cfa1b9bdef05cc04304e6739079e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR