Hydroxyurea ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A10831.03 A10831.06 A10831.14
A10831.03EA 28.6 EUR
A10831.03 A10831.06 A10831.14
Hydroxyurea ≥98%
Hydroxyurea

Purity: 98% CAS No.: 127-07-1 Molecular Formula: CH4N2O2 Formula Weight: 76.06 Melting Point: ca 140° dec. Beilstein Reference: 1741548 Merck Reference: 14,4848 MDL No.: MFCD00007943


An inhibitor of DNA synthesis and an anticancer agent

Formule: CH₄N₂O₂
Kookpunt: 296 °C (1013 hPa)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007943
CAS nummer: 127-07-1
EINECS: 204-821-7
Merck Index: 14,4848

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR