Benzenesulphonyl chloride ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Benzene sulphochloride

A10849.0B A10849.0I A10849.30
A10849.0BEA 66.1 EUR
A10849.0B A10849.0I A10849.30
Benzenesulphonyl chloride ≥98%
Benzenesulphonyl chloride

Purity: 98% CAS No.: 98-09-9 Molecular Formula: C6H5ClO2S Formula Weight: 176.62 Flash Point: 132°(270°F) Refractive Index: 1.5525 Odour: Irritating Melting Point: 14-15° Boiling Point: 252° Beilstein Reference: 606926 Merck Reference: 14,1072 UN No.: 2225 MDL No.: MFCD00007426

Formule: C₆H₅ClO₂S
MW: 176.62 g/mol
Kookpunt: 252 °C
Smeltpunt: 14…15 °C
Dichtheid: 1.377
Vlampunt: 132 °C (270 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007426
CAS nummer: 98-09-9
EINECS: 202-636-6
UN: 2225
ADR: 8 III
Merck Index: 13,01072

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR