Methyl toluene-4-sulphonate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Methyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid methyl ester, Methyl p-toluenesulphonate, Toluene-4-sulphonic acid methyl ester

A10881.0B A10881.30
A10881.0BEA 111 EUR
A10881.0B A10881.30
Methyl toluene-4-sulphonate ≥98%
Methyl toluene-4-sulphonate

Purity: 98% CAS No.: 80-48-8 Molecular Formula: C8H10O3S Formula Weight: 186.23 Storage Temperature: 2°C to 8°C Flash Point: 152°(305°F) Refractive Index: 1.5185 Melting Point: 24-28° Boiling Point: 140°/20mm Beilstein Reference: 609209 UN No.: 3261 MDL No.: MFCD00008417

Formule: C₈H₁₀O₃S
MW: 186.23 g/mol
Kookpunt: 140 °C (20 mmHg)
Smeltpunt: 24…28 °C
Dichtheid: 1.234
Vlampunt: 152 °C (305 °F)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008417
CAS nummer: 80-48-8
EINECS: 201-283-5
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR