DL-Camphor 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A10936.0C A10936.0E A10936.22 A10936.36
A10936.0CEA 392 EUR
A10936.0C A10936.0E A10936.22 A10936.36
DL-Camphor 96%
DL-Camphor

Purity: 96% CAS No.: 76-22-2 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 64°(147°F) Physical Form: Crystalline powder or lumps Density: 0.992 Appearance: White Odour: Camphor-like Melting Point: 172-176° Boiling Point: 204° Solubility: Soluble in alcohols, ether, chloroform, benzene Beilstein Reference: 1907611 Merck Reference: 14,1732 UN No.: 2717 MDL No.: MFCD00074738

Formule: C₁₀H₁₆O
Kookpunt: 204 °C
Smeltpunt: 172…176 °C
Dichtheid: 0,992
Vlampunt: 64 °C (147 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00074738
CAS nummer: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III
Merck Index: 14,1732

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR