1,1,2,2-Tetrafluoro-1,2-diiodoethane 96% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Stabilisatie: Stabilised with copper

A10954.06
A10954.06EA 162 EUR
A10954.06
1,1,2,2-Tetrafluoro-1,2-diiodoethane 96% stabilised
1,1,2,2-Tetrafluoro-1,2-diiodoethane

Stabilized with copper


Purity: 96% CAS No.: 354-65-4 Molecular Formula: IF2CCF2I Formula Weight: 353.82 Flash Point: None Refractive Index: 1.4950 Melting Point: -21° Boiling Point: 112-113° Beilstein Reference: 1740350 MDL No.: MFCD00039401

Formule: C₂F₄I₂
Kookpunt: 112…113 °C
Smeltpunt: –21 °C
Dichtheid: 2,629
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00039401
CAS nummer: 354-65-4
EINECS: 206-567-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR