6-Bromohexanoyl chloride ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A11063.06 A11063.14 A11063.22
A11063.06EA 40.6 EUR
A11063.06 A11063.14 A11063.22
6-Bromohexanoyl chloride ≥97%
6-Bromohexanoyl chloride

Purity: 97% CAS No.: 22809-37-6 Molecular Formula: C6H10BrClO Formula Weight: 213.50 Flash Point: 110°(230°F) Refractive Index: 1.4875 Odour: Acrid Boiling Point: 119-120°/9mm Beilstein Reference: 1749739 UN No.: 3265 MDL No.: MFCD00000761

Formule: BrH₂C(CH₂)₄COOCl
Kookpunt: 119…120 °C (9 mmHg)
Dichtheid: 1,397
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000761
CAS nummer: 22809-37-6
EINECS: 245-236-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR