DL-Lysine monohydrochloride ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A11066.14 A11066.22
A11066.14EA 38 EUR
A11066.14 A11066.22
DL-Lysine monohydrochloride ≥99%
DL-Lysine monohydrochloride

Purity: 99% CAS No.: 70-53-1 Molecular Formula: C6H14N2O2.HCl Formula Weight: 182.65 Melting Point: 267° dec. Beilstein Reference: 4711993

Formule: C₆H₁₅ClN₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064563
CAS nummer: 70-53-1
EINECS: 200-739-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR