D-(+)-Glucose monohydrate

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate

A11090.36 A11090.0E
A11090.36EA 24.5 EUR
A11090.36 A11090.0E
D-(+)-Glucose monohydrate
D-(+)-Glucose monohydrate

CAS No.: 14431-43-7 Molecular Formula: C6H12O6.H2O Formula Weight: 198.18 (180.16anhy) Physical Form: Cryst powder Appearance: White Melting Point: ca 83° MDL No.: MFCD00149450

Formule: C₆H₁₂O₆·H₂O
MW: 198.17 g/mol
Smeltpunt: ∼ 83 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149450
CAS nummer: 14431-43-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR