18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A11249.09 A11249.18 A11249.30
A11249.09EA 51.7 EUR
A11249.09 A11249.18 A11249.30
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99%
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)

Purity: 99% CAS No.: 17455-13-9 Molecular Formula: C12H24O6 Formula Weight: 264.32 Flash Point: 109°(228°F) Odour: Odourless Boiling Point: 116°/0.2mm Beilstein Reference: 1619616 Merck Reference: 14,2602 MDL No.: MFCD00005113

Formule: C₁₂H₂₄O₆
Kookpunt: 116 °C (0,2 mmHg)
Smeltpunt: 37…41 °C
Dichtheid: 1,175
Vlampunt: 109 °C (228 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005113
CAS nummer: 17455-13-9
EINECS: 241-473-5
Merck Index: 14,2602

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR