Koper(II)sulfaat pentahydraat ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A11262.0B A11262.0G A11262.0I
A11262.0BEA 42.5 EUR
A11262.0B A11262.0G A11262.0I
Koper(II)sulfaat pentahydraat ≥99%
Koper(II)sulfaat pentahydraat

Purity: 99% CAS No.: 7758-99-8 Molecular Formula: CuSO4.5H2O Formula Weight: 249.68 (159.60anhy) Odour: Odourless pH: 4 Merck Reference: 14,2653 UN No.: 3288 MDL No.: MFCD00149681

Formule: CuSO₄·5H₂O
Dichtheid: 2,270
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149681
CAS nummer: 7758-99-8
EINECS: 231-847-6
UN: 3288
ADR: 6.1,III
Bereik: 01-2119520566-40
Merck Index: 14,2653

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR