L-Alanine ethyl ester hydrochloride ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A11271.06 A11271.14
A11271.06EA 39.8 EUR
A11271.06 A11271.14
L-Alanine ethyl ester hydrochloride ≥98%
L-Alanine ethyl ester hydrochloride

Purity: 98+% CAS No.: 1115-59-9 Molecular Formula: C5H11NO2.HCl Formula Weight: 153.61 Melting Point: 74-78° Beilstein Reference: 3594395 Optical Rotation: +2.9° (c=2.5 in water) MDL No.: MFCD00063662

Formule: C₅H₁₁NO₂
Smeltpunt: 74…78 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00063662
CAS nummer: 1115-59-9
EINECS: 214-225-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR