Guanosine ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

A11328.18 A11328.14 A11328.22 A11328.30
A11328.18EA 95.6 EUR
A11328.18 A11328.14 A11328.22 A11328.30
Guanosine ≥98%
Guanosine

Purity: 98+% CAS No.: 118-00-3 Molecular Formula: C10H13N5O5 Formula Weight: 283.24 Melting Point: ca 250° dec. Beilstein Reference: 625911 Merck Reference: 14,4566 Optical Rotation: -72° (c=1.5 in 0.1N NaOH) MDL No.: MFCD00010182

Formule: C₁₀H₁₃N₅O₅
MW: 283.24 g/mol
Smeltpunt: ∼ 250 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010182
CAS nummer: 118-00-3
EINECS: 204-227-8
Merck Index: 14,4566

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR