Ethyl cyanoacetate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A11498.0E A11498.36
A11498.0EEA 171 EUR
A11498.0E A11498.36
Ethyl cyanoacetate ≥98%
Ethyl cyanoacetate

Purity: 98+% CAS No.: 105-56-6 Molecular Formula: C5H7NO2 Formula Weight: 113.12 Flash Point: 109°(228°F) Refractive Index: 1.4180 Odour: Weak Melting Point: -23° Boiling Point: 208-210° Beilstein Reference: 605871 Merck Reference: 14,3786 MDL No.: MFCD00001940

Formule: C₅H₇NO₂
Kookpunt: 208…210 °C
Smeltpunt: –23 °C
Dichtheid: 1,063
Vlampunt: 109 °C (228 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001940
CAS nummer: 105-56-6
EINECS: 203-309-0
Merck Index: 14,3786

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR