4-Methyl-2-pentaanon ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK

A11618.0F A11618.AP
A11618.0FEA 66.4 EUR
A11618.0F A11618.AP
4-Methyl-2-pentaanon ≥99%
4-Methyl-2-pentaanon

Purity: 99% CAS No.: 108-10-1 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 13°(55°F) Refractive Index: 1.3960 Odour: Mild Melting Point: -84° Boiling Point: 117-118° Beilstein Reference: 605399 Merck Reference: 14,5207 UN No.: 1245 MDL No.: MFCD00008938

Formule: (CH₃)₂CHCH₂COCH₃
MW: 100.16 g/mol
Kookpunt: 117…118 °C
Smeltpunt: –84 °C
Dichtheid: 0.801
Vlampunt: 13 °C (55 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008938
CAS nummer: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3 II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR