Pentafluorobenzyl bromide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A11623.14 A11623.06 A11623.22
A11623.14EA 182 EUR
A11623.14 A11623.06 A11623.22
Pentafluorobenzyl bromide ≥98%
Pentafluorobenzyl bromide

Purity: 98+% CAS No.: 1765-40-8 Molecular Formula: C7H2BrF5 Formula Weight: 261.00 Flash Point: 83°(181°F) Refractive Index: 1.4720 Melting Point: 18-20° Boiling Point: 174-175° Beilstein Reference: 647808 UN No.: 3265 MDL No.: MFCD00000299

Formule: (C₆F₅)CH₂Br
Kookpunt: 174…175 °C
Smeltpunt: 18…20 °C
Dichtheid: 1,844
Vlampunt: 83 °C (181 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000299
CAS nummer: 1765-40-8
EINECS: 217-182-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR