4-Nitrobenzaldehyde ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A11655.14 A11655.22 A11655.36
A11655.14EA 35.4 EUR
A11655.14 A11655.22 A11655.36
4-Nitrobenzaldehyde ≥99%
4-Nitrobenzaldehyde

Purity: 99% CAS No.: 555-16-8 Molecular Formula: C7H5NO3 Formula Weight: 151.12 Melting Point: 104-106° Beilstein Reference: 386796 Merck Reference: 14,6587 MDL No.: MFCD00007346

Formule: C₇H₅NO₃
Kookpunt: 300 °C (1013 hPa)
Smeltpunt: 104…106 °C
Dichtheid: 1,546 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007346
CAS nummer: 555-16-8
EINECS: 209-084-5
Merck Index: 14,6587

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR