Potassium chloride ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A11662.0B A11662.0G A11662.0I
A11662.0BEA 40.1 EUR
A11662.0B A11662.0G A11662.0I
Potassium chloride ≥99%
Potassium chloride

Purity: 99% CAS No.: 7447-40-7 Molecular Formula: KCl Formula Weight: 74.55 Refractive Index: 1.490 Odour: Odourless Boiling Point: 1500° subl. Merck Reference: 14,7621 MDL No.: MFCD00011360

Formule: KCl
Kookpunt: 1500 °C (sublimatie)
Smeltpunt: 773 °C
Dichtheid: 1,984
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,7621

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR