Aminoiminomethanesulphinic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A11885.22
A11885.22EA 37.1 EUR
A11885.22
Aminoiminomethanesulphinic acid ≥98%
Aminoiminomethanesulphinic acid
Formule: CH₄N₂O₂S
Dichtheid: 1,68
MDL Nummer: MFCD00002397
CAS nummer: 1758-73-2
UN: 3341
ADR: 4.2,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR