Pyridoxine hydrochloride ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A12041.14 A12041.22 A12041.36
A12041.14EA 39.2 EUR
A12041.14 A12041.22 A12041.36
Pyridoxine hydrochloride ≥99%
Pyridoxine hydrochloride

Purity: 99% CAS No.: 58-56-0 Molecular Formula: C8H11NO3.HCl Formula Weight: 205.64 Odour: Odourless pH: 3 Melting Point: ca 206° dec. Beilstein Reference: 3632435 Merck Reference: 14,7982 MDL No.: MFCD00012807

Formule: C₈H₁₂ClNO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012807
CAS nummer: 58-56-0
EINECS: 200-386-2
Merck Index: 14,7982

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR