2-Phenylbenzoic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A12049.06 A12049.14 A12049.22
A12049.06EA 21.7 EUR
A12049.06 A12049.14 A12049.22
2-Phenylbenzoic acid ≥98%
2-Phenylbenzoic acid

Purity: 98% CAS No.: 947-84-2 Molecular Formula: C13H10O2 Formula Weight: 198.22 Odour: Odourless Melting Point: 110-114° Boiling Point: 343-344° Beilstein Reference: 974075 MDL No.: MFCD00002463

Formule: C₁₃H₁₀O₂
Kookpunt: 343…344 °C
Smeltpunt: 110…114 °C
Dichtheid: 1,458 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002463
CAS nummer: 947-84-2
EINECS: 213-432-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR