Diethyl methylmalonate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Diethyl 2-methylmalonate

A12078.0B A12078.18 A12078.30
A12078.0BEA 277 EUR
A12078.0B A12078.18 A12078.30
Diethyl methylmalonate ≥99%
Diethyl methylmalonate

Purity: 99% CAS No.: 609-08-5 Molecular Formula: C8H14O4 Formula Weight: 174.20 Flash Point: 85°(185°F) Refractive Index: 1.4130 Boiling Point: 198-199° Beilstein Reference: 637028 MDL No.: MFCD00009162

Formule: C₈H₁₄O₄
MW: 174.2 g/mol
Kookpunt: 198…199 °C
Dichtheid: 1.02
Vlampunt: 85 °C (185 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009162
CAS nummer: 609-08-5
EINECS: 210-175-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR