2,3,4,5,6-Pentafluorobenzyl alcohol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A12119.06 A12119.14
A12119.06EA 43.5 EUR
A12119.06 A12119.14
2,3,4,5,6-Pentafluorobenzyl alcohol ≥98%
2,3,4,5,6-Pentafluorobenzyl alcohol

Purity: 98% CAS No.: 440-60-8 Molecular Formula: C7H3F5O Formula Weight: 198.09 Flash Point: 87°(188°F) Melting Point: 32-36° Boiling Point: 182° Beilstein Reference: 2052669 MDL No.: MFCD00004602

Formule: C₇H₃F₅O
Kookpunt: 182 °C
Smeltpunt: 32…36 °C
Vlampunt: 87 °C (188 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004602
CAS nummer: 440-60-8
EINECS: 207-126-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR