Toluene-4-sulphonamide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: PTSA, p-Toluenesulphonamide, 4-Methylbenzenesulphonamide, 4-Toluenesulphonamide

A12156.0C A12156.0E A12156.30 A12156.36
A12156.0CEA 438 EUR
A12156.0C A12156.0E A12156.30 A12156.36
Toluene-4-sulphonamide ≥98%
Toluene-4-sulphonamide

Purity: 98+% CAS No.: 70-55-3 Molecular Formula: C7H9NO2S Formula Weight: 171.22 Flash Point: 202°(395°F) Melting Point: 136-139° Boiling Point: 221°/10mm Beilstein Reference: 472689 MDL No.: MFCD00011692

Formule: C₇H₉NO₂S
MW: 171.22 g/mol
Kookpunt: 221 °C (10 mmHg)
Smeltpunt: 136…139 °C
Vlampunt: 202 °C (395 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011692
CAS nummer: 70-55-3
EINECS: 200-741-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR