Sulphur trioxide pyridine complex 98% active SO₃ ∼ 48-50%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A12202.09 A12202.0B A12202.18 A12202.30
A12202.09EA 26.5 EUR
A12202.09 A12202.0B A12202.18 A12202.30
Sulphur trioxide pyridine complex 98% active SO₃ ∼ 48-50%
Sulphur trioxide pyridine complex

Purity: 98% CAS No.: 26412-87-3 Molecular Formula: C5H5NO3S Formula Weight: 159.16 Melting Point: ca 155-165° Beilstein Reference: 3704116 UN No.: 3261 MDL No.: MFCD00012437

Formule: C₅H₅NO₃S
MW: 159.16 g/mol
Smeltpunt: ∼ 155…165 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012437
CAS nummer: 26412-87-3
EINECS: 247-683-3
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR