4-(4-Nitrophenylazo)resorcinol ≥90%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A12232.14 A12232.22 A12232.36
A12232.14EA 42.2 EUR
A12232.14 A12232.22 A12232.36
4-(4-Nitrophenylazo)resorcinol ≥90%
4-(4-Nitrophenylazo)resorcinol

Purity: 95% CAS No.: 74-39-5 Molecular Formula: C12H9N3O4 Formula Weight: 259.22 Melting Point: ca 185° dec. Beilstein Reference: 674709 Merck Reference: 14,5695 MDL No.: MFCD00007310

Formule: C₁₂H₉N₃O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007310
CAS nummer: 74-39-5
EINECS: 200-808-5
Merck Index: 14,5695

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR