2,3-Dichloropyridine ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A12241.06 A12241.14 A12241.22
A12241.06EA 38.4 EUR
A12241.06 A12241.14 A12241.22
2,3-Dichloropyridine ≥99%
2,3-Dichloropyridine

Purity: 99% CAS No.: 2402-77-9 Molecular Formula: C5H3Cl2N Formula Weight: 147.99 Odour: Unpleasant Melting Point: 65-69° Beilstein Reference: 109811 UN No.: 2811 MDL No.: MFCD00006229

Formule: C₅H₃Cl₂N
Kookpunt: 202,5…203.5 °C (1013 hPa)
Smeltpunt: 65…69 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006229
CAS nummer: 2402-77-9
EINECS: 219-281-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR