Dibenzothiophene ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A12288.14 A12288.22 A12288.36
A12288.14EA 39.4 EUR
A12288.14 A12288.22 A12288.36
Dibenzothiophene ≥98%
Dibenzothiophene

Purity: 98% CAS No.: 132-65-0 Molecular Formula: C12H8S Formula Weight: 184.25 Flash Point: 170°(338°F) Melting Point: 97-100° Boiling Point: 332-333° Beilstein Reference: 121101 UN No.: 3077 MDL No.: MFCD00004969

Formule: C₁₂H₈S
Kookpunt: 332…333 °C
Smeltpunt: 97…100 °C
Dichtheid: 1,1295 g/cm³ (125 °C)
Vlampunt: 170 °C (338 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004969
CAS nummer: 132-65-0
EINECS: 205-072-9
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR