Phosphoenolpyruvic acid monocyclohexylammonium salt 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A12385.03 A12385.14 A12385.06
A12385.03EA 82 EUR
A12385.03 A12385.14 A12385.06
Phosphoenolpyruvic acid monocyclohexylammonium salt 98%
Phosphoenolpyruvic acid monocyclohexylammonium salt

Purity: 98% CAS No.: 10526-80-4 Molecular Formula: C9H18NO6P Formula Weight: 267.22 Storage Temperature: -30°C to -10°C Melting Point: ca 140° dec. MDL No.: MFCD00036375

Formule: C₃H₅O₆P
MDL Nummer: MFCD00036375
CAS nummer: 10526-80-4
EINECS: 234-084-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR