Methyl-α-D-glucopyranoside ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: α-D-Methylglucoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

A12484.22 A12484.36
A12484.22EA 32 EUR
A12484.22 A12484.36
Methyl-α-D-glucopyranoside ≥98%
Methyl-α-D-glucopyranoside

Purity: 98+% CAS No.: 97-30-3 Molecular Formula: C7H14O6 Formula Weight: 194.18 Melting Point: 166-171° Solubility: Soluble in water. Insoluble in ether Beilstein Reference: 81568 Merck Reference: 14,6080 Optical Rotation: +159° (c=10 in water) MDL No.: MFCD00064086

Formule: C₇H₁₄O₆
MW: 194.18 g/mol
Smeltpunt: 166…171 °C
Dichtheid: 1.460
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064086
CAS nummer: 97-30-3
EINECS: 202-571-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR