Methyl laurate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A12492.06 A12492.14 A12492.22
A12492.06EA 42 EUR
A12492.06 A12492.14 A12492.22
Methyl laurate ≥99%
Methyl laurate

Purity: 99% CAS No.: 111-82-0 Molecular Formula: CH3(CH2)10CO2CH3 Formula Weight: 214.35 Flash Point: 125°(257°F) Refractive Index: 1.4325 Melting Point: 4-5° Boiling Point: 261-262° Beilstein Reference: 1767780 MDL No.: MFCD00008966

Formule: C₁₃H₂₆O₂
Kookpunt: 261…262 °C
Smeltpunt: 4…5 °C
Dichtheid: 0,870
Vlampunt: 125 °C (257 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008966
CAS nummer: 111-82-0
EINECS: 203-911-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR