Phthalimide ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A12584.0C A12584.0E A12584.36
A12584.0CEA 304 EUR
A12584.0C A12584.0E A12584.36
Phthalimide ≥99%
Phthalimide

Purity: 99% CAS No.: 85-41-6 Molecular Formula: C8H5NO2 Formula Weight: 147.14 Flash Point: 165°(329°F) Odour: Odourless pH: 3.8 Melting Point: 232-236° Boiling Point: 366° Solubility: Slightly soluble in water. Soluble in aqueous alkali hydroxides Beilstein Reference: 118522 Merck Reference: 14,7373 MDL No.: MFCD00005881

Formule: C₈H₅NO₂
Kookpunt: 366 °C
Smeltpunt: 232…236 °C
Dichtheid: 1,21
Vlampunt: 165 °C (329 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005881
CAS nummer: 85-41-6
EINECS: 201-603-3
Merck Index: 14,7373

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR