1-Phenylpiperazine ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A12610.14 A12610.22 A12610.36
A12610.14EA 32.2 EUR
A12610.14 A12610.22 A12610.36
1-Phenylpiperazine ≥98%
1-Phenylpiperazine

Purity: 98+% CAS No.: 92-54-6 Molecular Formula: C10H14N2 Formula Weight: 162.24 Flash Point: 141°(286°F) Refractive Index: 1.5880 Melting Point: 17-19° Boiling Point: 285-286° Beilstein Reference: 132157 UN No.: 2927 MDL No.: MFCD00005957

Formule: C₁₀H₁₄N₂
Kookpunt: 285…286 °C
Smeltpunt: 17…19 °C
Dichtheid: 1,064
Vlampunt: 141 °C (286 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005957
CAS nummer: 92-54-6
EINECS: 202-165-6
UN: 2927
ADR: 6.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR